Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC42EP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145057.3 | NP_659494.2 | 3 | 816 | ||
GenBank mRNA | BC024327.1 | - | 3 | 787 | 4390710 | |
BC065910.1 | - | 3 | 721 | 6572564 |