Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152486.2 | NP_689699.2 | 7 | 668 | ||
GenBank mRNA | AK054643.1 | - | 1 | 70 | ||
BC024295.2 | - | 3 | 291 | 3354502 | ||
BC033213.1 | - | 1 | 33 | 5014368 | ||
CN293867.1 | - | 4 | 308 | |||
JX093066.1 | - | 6 | 591 | |||
JX093068.1 | - | 6 | 591 | |||
JX093069.1 | - | 6 | 588 | |||
JX093070.1 | - | 6 | 591 | |||
JX093071.1 | - | 6 | 588 | |||
JX093072.1 | - | 5 | 540 | |||
JX093073.1 | - | 6 | 591 | |||
JX093074.1 | - | 6 | 558 | |||
JX093075.1 | - | 6 | 591 | |||
JX093077.1 | - | 6 | 588 | |||
JX093078.1 | - | 6 | 588 | |||
JX093079.1 | - | 6 | 591 | |||
JX093080.1 | - | 6 | 591 | |||
JX093081.1 | - | 6 | 588 | |||
JX093082.1 | - | 6 | 588 | |||
JX093083.1 | - | 6 | 588 | |||
JX093084.1 | - | 6 | 591 | |||
JX093085.1 | - | 6 | 591 | |||
JX093086.1 | - | 6 | 591 | |||
JX093087.1 | - | 6 | 588 | |||
JX093088.1 | - | 6 | 588 | |||
JX093089.1 | - | 6 | 588 | |||
JX093090.1 | - | 6 | 591 | |||
JX093091.1 | - | 4 | 415 | |||
JX093092.1 | - | 6 | 591 | |||
JX093093.1 | - | 6 | 588 | |||
JX093094.1 | - | 6 | 591 | |||
JX093095.1 | - | 6 | 588 | |||
JX093099.1 | - | 7 | 676 | |||
JX093100.1 | - | 7 | 673 | |||
JX093101.1 | - | 7 | 676 | |||
JX093103.1 | - | 6 | 625 | |||
JX093104.1 | - | 5 | 463 | |||
JX093105.1 | - | 5 | 466 | |||
JX093106.1 | - | 5 | 463 | |||
JX093107.1 | - | 5 | 463 | |||
JX093108.1 | - | 5 | 463 | |||
JX093109.1 | - | 4 | 355 | |||
JX093110.1 | - | 4 | 358 |