Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YDJC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017964.1 | NP_001017964.1 | 5 | 1024 | ||
XM_011529906.2 | XP_011528208.1 | 4 | 1121 | |||
GenBank mRNA | BC096754.1 | - | 5 | 1097 | ||
BC110075.1 | - | 4 | 830 | |||
BC110799.1 | - | 5 | 1068 | |||
BC133043.1 | - | 5 | 1178 | |||
BC133047.1 | - | 5 | 1178 |