Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148961.3 | NP_683764.1 | 4 | 416 | ||
XM_017003409.1 | XP_016858898.1 | 4 | 370 | |||
XM_017003410.1 | XP_016858899.1 | 4 | 1051 | |||
GenBank mRNA | BC105085.1 | - | 3 | 304 | 8144088 | |
BC105087.1 | - | 3 | 304 | 8144090 |