Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAHD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320848.1 | NP_001307777.1 | 6 | 899 | ||
NM_199336.2 | NP_955368.1 | 5 | 773 | |||
XM_011510745.2 | XP_011509047.1 | 6 | 889 | |||
XM_011510746.2 | XP_011509048.1 | 6 | 1008 | |||
XM_011510747.2 | XP_011509049.1 | 5 | 773 | |||
XM_011510748.2 | XP_011509050.1 | 6 | 889 | |||
XM_017003470.1 | XP_016858959.1 | 6 | 886 | |||
XM_017003471.1 | XP_016858960.1 | 6 | 889 | |||
XM_017003472.1 | XP_016858961.1 | 5 | 822 | |||
GenBank mRNA | AL833869.1 | - | 4 | 573 | ||
BC064511.1 | - | 5 | 736 | 6644987 |
Gene Symbol: LOC107984110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017005470.1 | XP_016860959.1 | 4 | 1704 |