Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAHD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320848.1 | NP_001307777.1 | 3 | 502 | ||
NM_001320849.1 | NP_001307778.1 | 2 | 376 | |||
NM_199336.2 | NP_955368.1 | 2 | 376 | |||
XM_011510745.2 | XP_011509047.1 | 3 | 492 | |||
XM_011510746.2 | XP_011509048.1 | 3 | 611 | |||
XM_011510747.2 | XP_011509049.1 | 2 | 376 | |||
XM_011510748.2 | XP_011509050.1 | 3 | 492 | |||
XM_011510750.2 | XP_011509052.1 | 3 | 494 | |||
XM_017003470.1 | XP_016858959.1 | 3 | 489 | |||
XM_017003471.1 | XP_016858960.1 | 3 | 492 | |||
XM_017003472.1 | XP_016858961.1 | 2 | 368 | |||
XM_017003473.1 | XP_016858962.1 | 2 | 376 | |||
GenBank mRNA | AK295056.1 | - | 2 | 329 | ||
AL833869.1 | - | 1 | 176 | |||
BC064511.1 | - | 2 | 339 | 6644987 | ||
BI547630.1 | - | 2 | 381 |