Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024675.1 | NP_001019846.1 | 1 | 1235 | ||
GenBank mRNA | BC137111.1 | - | 1 | 1215 | 9021108 | |
BC137114.1 | - | 1 | 1215 | 9021111 |