Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGAM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170543.1 | NP_001164014.1 | 3, 4 | 571 | ||
NM_001170544.1 | NP_001164015.1 | 3, 4 | 571 | |||
NM_138575.3 | NP_612642.2 | 3, 4 | 571 | |||
GenBank mRNA | AK097688.1 | - | 3, 4 | 918 | ||
BC008196.1 | - | 3, 4 | 528 | 3048106 | ||
BC047906.2 | - | 2, 3 | 226 | |||
DR155760.1 | - | 3, 4 | 539 | |||
EU249757.1 | - | 3, 4 | 559 |