Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015050.2 | NP_001015050.2 | 2, 3 | 277 | ||
NM_001195223.1 | NP_001182152.1 | 3, 4 | 388 | |||
XM_017005844.1 | XP_016861333.1 | 3, 4 | 3652 | |||
XM_017005845.1 | XP_016861334.1 | 3, 4 | 278 | |||
XM_017005846.1 | XP_016861335.1 | 2, 3 | 3541 | |||
XM_017005847.1 | XP_016861336.1 | 2, 3 | 167 | |||
GenBank mRNA | BC073816.1 | - | 2, 3 | 272 | 5419254 | |
BI085096.1 | - | 3, 4 | 380 | |||
CD685738.1 | - | 2, 3 | 298 |