Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TVP23C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135036.1 | NP_001128508.1 | 3 | 380 | ||
NM_145301.2 | NP_660344.2 | 3 | 380 | |||
GenBank mRNA | AK310363.1 | - | 3 | 339 | ||
BC011952.1 | - | 3 | 280 | 3860433 |
Gene Symbol: TVP23C-CDRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204478.1 | NP_001191407.1 | 3 | 380 | ||
GenBank mRNA | AB074181.1 | - | 3 | 310 |