Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174922.4 | NP_777582.4 | 10, 11 | 1180 | ||
XM_006716527.2 | XP_006716590.1 | 10, 11 | 1170 | |||
XM_011516907.2 | XP_011515209.1 | 10, 11 | 1893 | |||
XM_011516909.1 | XP_011515211.1 | 10, 11 | 1180 | |||
XM_011516910.1 | XP_011515212.1 | 11, 12 | 1203 | |||
XM_017013174.1 | XP_016868663.1 | 10, 11 | 1218 | |||
GenBank mRNA | AK092773.1 | - | 10, 11 | 1143 | ||
BC064345.1 | - | 1, 2 | 131 | |||
BC085013.1 | - | 5, 6 | 490 | |||
BC101659.1 | - | 10, 11 | 1120 | 8069165 | ||
BC143326.1 | - | 9, 10 | 947 | |||
BU845476.1 | - | 5, 6 | 488 |