Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005277637.2 | XP_005277694.1 | 9, 10 | 871 | ||
XM_005277640.2 | XP_005277697.1 | 8, 9 | 784 |
Gene Symbol: CBWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145356.1 | NP_001138828.1 | 9, 10 | 811 | ||
XM_005251515.1 | XP_005251572.1 | 8, 9 | 724 | |||
XM_011517961.2 | XP_011516263.1 | 10, 11 | 903 | |||
GenBank mRNA | AK297292.1 | - | 10, 11 | 850 |
Gene Symbol: CBWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005263600.2 | XP_005263657.1 | 9, 10 | 883 | ||
XM_005263604.2 | XP_005263661.1 | 8, 9 | 796 | |||
XM_005263605.3 | XP_005263662.1 | 10, 11 | 918 |
Gene Symbol: CBWD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024916.3 | NP_001020087.2 | 8, 9 | 761 | ||
XM_005272749.1 | XP_005272806.1 | 9, 10 | 808 | |||
GenBank mRNA | BC082271.1 | - | 8, 9 | 657 |
Gene Symbol: CBWD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015021.1 | XP_016870510.1 | 8, 9 | 811 | ||
XM_017015023.1 | XP_016870512.1 | 8, 9 | 811 | |||
XM_017015025.1 | XP_016870514.1 | 7, 8 | 724 | |||
XM_017015030.1 | XP_016870519.1 | 8, 9 | 811 | |||
XM_017015035.1 | XP_016870524.1 | 9, 10 | 871 | |||
XM_017015036.1 | XP_016870525.1 | 8, 9 | 784 |