Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008703.2 | NP_001008703.1 | 5 | 880 | ||
NM_001008704.2 | NP_001008704.1 | 5 | 820 | |||
NM_001287396.1 | NP_001274325.1 | 5 | 1072 | |||
NM_001287397.1 | NP_001274326.1 | 4 | 1106 | |||
NM_001287398.1 | NP_001274327.1 | 4 | 854 | |||
NM_178508.4 | NP_848603.2 | 5 | 1132 | |||
GenBank mRNA | AK290164.1 | - | 5 | 814 | ||
BC023627.2 | - | 5 | 752 | 4764944 | ||
BC047919.1 | - | 5 | 1013 | 6190218 | ||
BM474526.1 | - | 4 | 772 | |||
BU956518.1 | - | 4 | 619 |