Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTAG1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139250.1 | NP_640343.1 | 1 | 274 |
Gene Symbol: CTAG1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001327.2 | NP_001318.1 | 1 | 274 | ||
GenBank mRNA | AF038567.1 | - | 1 | 309 | ||
AJ003149.1 | - | 1 | 271 | |||
AJ275978.1 | - | 1 | 143 | |||
BC130362.1 | - | 1 | 278 | 40146393 | ||
BC130364.1 | - | 1 | 278 | 40146395 | ||
U87459.1 | - | 1 | 274 |