Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4E1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099408.1 | NP_001092878.1 | 6 | 900 | ||
XM_011534499.2 | XP_011532801.1 | 6 | 981 | |||
GenBank mRNA | AK094270.1 | - | 5 | 1268 | ||
DR008159.1 | - | 3 | 131 | |||
DR731203.1 | - | 3 | 137 |