Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAMSTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130915.1 | NP_001124387.1 | 6 | 555 | ||
NM_001297753.1 | NP_001284682.1 | 4 | 863 | |||
NM_182574.2 | NP_872380.1 | 4 | 799 | |||
XM_011526807.2 | XP_011525109.1 | 4 | 804 | |||
XM_011526808.2 | XP_011525110.1 | 6 | 923 | |||
XM_011526809.2 | XP_011525111.1 | 4 | 797 | |||
XM_017026640.1 | XP_016882129.1 | 5 | 1323 | |||
XM_017026641.1 | XP_016882130.1 | 5 | 1284 | |||
XM_017026642.1 | XP_016882131.1 | 4 | 827 | |||
XM_017026643.1 | XP_016882132.1 | 5 | 1164 | |||
XM_017026644.1 | XP_016882133.1 | 5 | 1134 | |||
XM_017026645.1 | XP_016882134.1 | 3 | 764 | |||
GenBank mRNA | AK093389.1 | - | 4 | 799 | ||
AK131427.1 | - | 6 | 562 | |||
BC105056.1 | - | 2 | 223 | 8144059 | ||
BC105058.1 | - | 2 | 223 | 8144061 | ||
BC106047.1 | - | 4 | 863 | 5786639 | ||
BC143443.1 | - | 2 | 223 | 9051951 |