Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR10H5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004466.1 | NP_001004466.1 | 1 | 782 | ||
GenBank mRNA | BC136869.1 | - | 1 | 880 | 9020860 |
Gene Symbol: OR10H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013940.2 | NP_039228.1 | 1 | 871 | ||
XM_011527911.1 | XP_011526213.1 | 2 | 828 | |||
GenBank mRNA | BC130360.1 | - | 1 | 871 | 40146391 | |
BC132969.1 | - | 1 | 871 | 40146991 |
Set Membership: |
G-protein coupled receptor |