Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HUWE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031407.6 | NP_113584.3 | 23 | 2561 | ||
XM_005261965.3 | XP_005262022.1 | 22 | 2628 | |||
XM_011530747.1 | XP_011529049.1 | 24 | 2804 | |||
XM_011530751.2 | XP_011529053.1 | 23 | 2672 | |||
XM_017029191.1 | XP_016884680.1 | 22 | 5556 | |||
XM_017029192.1 | XP_016884681.1 | 22 | 5556 | |||
XM_017029193.1 | XP_016884682.1 | 22 | 5535 | |||
XM_017029194.1 | XP_016884683.1 | 22 | 5556 | |||
XM_017029195.1 | XP_016884684.1 | 22 | 5556 | |||
XM_017029196.1 | XP_016884685.1 | 22 | 5556 | |||
XM_017029197.1 | XP_016884686.1 | 22 | 5556 | |||
XM_017029198.1 | XP_016884687.1 | 23 | 2898 | |||
XM_017029199.1 | XP_016884688.1 | 24 | 2825 | |||
XM_017029200.1 | XP_016884689.1 | 23 | 2695 | |||
XM_017029201.1 | XP_016884690.1 | 23 | 2712 | |||
XM_017029202.1 | XP_016884691.1 | 23 | 2688 | |||
XM_017029203.1 | XP_016884692.1 | 23 | 2694 | |||
XM_017029204.1 | XP_016884693.1 | 21 | 5306 | |||
XM_017029205.1 | XP_016884694.1 | 21 | 5286 | |||
XM_017029206.1 | XP_016884695.1 | 22 | 5556 | |||
GenBank mRNA | AK056671.1 | - | 6 | 787 | ||
AK294575.1 | - | 10 | 1161 | |||
AY772009.1 | - | 21 | 2206 | |||
AY929612.1 | - | 20 | 2158 | |||
DQ097177.1 | - | 23 | 2560 |