Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320185.1 | NP_001307114.1 | 3 | 237 | ||
NM_181781.3 | NP_861446.2 | 3 | 237 | |||
NM_182975.3 | NP_892020.1 | 3 | 237 | |||
NM_182976.3 | NP_892021.1 | 3 | 237 | |||
XM_005270779.4 | XP_005270836.1 | 3 | 217 | |||
XM_011541290.2 | XP_011539592.1 | 3 | 232 | |||
GenBank mRNA | AK000410.1 | - | 3 | 204 | ||
AK297073.1 | - | 3 | 162 | |||
AK303707.1 | - | 3 | 164 | |||
BC014899.1 | - | 3 | 195 | |||
BC052645.1 | - | 3 | 211 | |||
BC053888.1 | - | 3 | 198 | |||
BC070341.1 | - | 3 | 181 | |||
BC090866.1 | - | 3 | 231 | 6503526 | ||
CN299618.1 | - | 3 | 247 | |||
DB111509.1 | - | 3 | 213 |