Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195001.1 | NP_001181930.1 | 24 | 3595 | ||
NM_005704.4 | NP_005695.3 | 25 | 3634 | |||
NM_133177.3 | NP_573438.3 | 25 | 3622 | |||
NM_133178.3 | NP_573439.2 | 24 | 3604 | |||
XM_006710269.2 | XP_006710332.1 | 25 | 3597 | |||
XM_016999992.1 | XP_016855481.1 | 26 | 3627 | |||
XM_016999993.1 | XP_016855482.1 | 26 | 3618 | |||
XM_016999994.1 | XP_016855483.1 | 25 | 3600 | |||
XM_016999995.1 | XP_016855484.1 | 21 | 2793 | |||
GenBank mRNA | AB208855.1 | - | 20 | 2898 | ||
BC146655.1 | - | 24 | 3562 | 40147934 | ||
U71075.1 | - | 25 | 3496 | |||
U73727.1 | - | 24 | 3595 | |||
X95712.1 | - | 26 | 3556 | |||
X97198.1 | - | 24 | 3589 |