Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AURKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323303.1 | NP_001310232.1 | 7 | 1132 | ||
NM_001323304.1 | NP_001310233.1 | 7 | 895 | |||
NM_001323305.1 | NP_001310234.1 | 8 | 1005 | |||
NM_003600.3 | NP_003591.2 | 6 | 1022 | |||
NM_198433.2 | NP_940835.1 | 8 | 1230 | |||
NM_198434.2 | NP_940836.1 | 7 | 921 | |||
NM_198435.2 | NP_940837.1 | 6 | 811 | |||
NM_198436.2 | NP_940838.1 | 7 | 907 | |||
NM_198437.2 | NP_940839.1 | 6 | 797 | |||
XM_017028034.1 | XP_016883523.1 | 8 | 1352 | |||
XM_017028035.1 | XP_016883524.1 | 6 | 712 | |||
GenBank mRNA | AF008551.1 | - | 5 | 622 | ||
AF011468.1 | - | 6 | 878 | |||
AK301769.1 | - | 6 | 484 | |||
AK312776.1 | - | 6 | 689 | |||
BC001280.1 | - | 7 | 754 | 3456017 | ||
BC002499.2 | - | 6 | 750 | 3051177 | ||
BC006423.1 | - | 6 | 647 | 3532438 | ||
BC027464.1 | - | 7 | 789 | 4449474 | ||
BQ435059.1 | - | 6 | 643 | |||
BU171303.1 | - | 6 | 743 | |||
D84212.1 | - | 7 | 788 |
Gene Symbol: AURKAPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001587.1 | 1 | 420 |