Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195001.1 | NP_001181930.1 | 2, 3 | 342 | ||
NM_005704.4 | NP_005695.3 | 2, 3 | 342 | |||
NM_133177.3 | NP_573438.3 | 2, 3 | 342 | |||
NM_133178.3 | NP_573439.2 | 2, 3 | 342 | |||
XM_006710269.2 | XP_006710332.1 | 2, 3 | 317 | |||
XM_016999992.1 | XP_016855481.1 | 2, 3 | 317 | |||
XM_016999993.1 | XP_016855482.1 | 2, 3 | 317 | |||
XM_016999994.1 | XP_016855483.1 | 2, 3 | 317 | |||
GenBank mRNA | BC146655.1 | - | 2, 3 | 309 | 40147934 | |
U71075.1 | - | 2, 3 | 219 | |||
U73727.1 | - | 2, 3 | 333 | |||
X95712.1 | - | 3, 4 | 273 | |||
X97198.1 | - | 2, 3 | 345 |