Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193635.1 | NP_001180564.1 | 7 | 674 | ||
NM_001193636.1 | NP_001180565.1 | 6 | 917 | |||
NM_001193637.1 | NP_001180566.1 | 4 | 794 | |||
NM_198083.3 | NP_932349.2 | 6 | 716 | |||
GenBank mRNA | AK301373.1 | - | 3 | 421 | ||
BC000663.2 | - | 6 | 624 | 3349226 | ||
DN237881.1 | - | 4 | 481 | |||
DQ088987.1 | - | 4 | 215 | |||
DQ088988.1 | - | 4 | 215 |