Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPIHBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178172.5 | NP_835466.2 | 4 | 487 | ||
GenBank mRNA | AY245915.1 | - | 4 | 487 | ||
BC035810.1 | - | 4 | 487 | 5754421 | ||
BC063857.1 | - | 4 | 415 | 6183386 |