Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFPL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145014.1 | NP_001138486.1 | 2, 3 | 340 | ||
XM_011526915.2 | XP_011525217.1 | 3, 4 | 1628 |