Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QRFP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198180.2 | NP_937823.1 | 3 | 880 | ||
GenBank mRNA | AB109625.1 | - | 1 | 359 | ||
BC101127.2 | - | 1 | 359 | 40014164 | ||
BC101128.1 | - | 1 | 359 | 40014167 |