Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005707.1 | NP_005698.1 | 3 | 1123 | ||
GenBank mRNA | AF083930.1 | - | 3 | 268 | ||
AK096970.1 | - | 3 | 1123 | |||
AK225816.1 | - | 3 | 1090 | |||
BC016992.2 | - | 1 | 53 | |||
BC092464.1 | - | 5 | 447 | |||
BC131704.1 | - | 4 | 425 |