Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005708.3 | NP_005699.1 | 8 | 2046 | ||
XM_011521044.2 | XP_011519346.1 | 8 | 1371 | |||
XM_017020298.1 | XP_016875787.1 | 8 | 3654 | |||
XM_017020299.1 | XP_016875788.1 | 8 | 2421 | |||
XM_017020300.1 | XP_016875789.1 | 8 | 1278 | |||
XM_017020301.1 | XP_016875790.1 | 7 | 1249 | |||
XM_017020302.1 | XP_016875791.1 | 5 | 1043 | |||
GenBank mRNA | AF105267.1 | - | 8 | 1999 | ||
AF111178.1 | - | 8 | 2048 | |||
AK074741.1 | - | 9 | 1500 | |||
AK290144.1 | - | 8 | 1567 | |||
AY358462.1 | - | 8 | 1979 | |||
BC106947.2 | - | 8 | 1894 | 40034564 |