Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMY1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006121.3 | NP_001006121.1 | 12 | 1657 | ||
NM_001320948.1 | NP_001307877.1 | 11 | 1546 | |||
XM_011531433.1 | XP_011529735.1 | 9 | 1284 | |||
XM_011531434.1 | XP_011529736.1 | 11 | 1456 | |||
XM_017030057.1 | XP_016885546.1 | 10 | 1322 | |||
GenBank mRNA | BC136862.1 | - | 11 | 1459 | 9020853 | |
BC144459.1 | - | 11 | 1459 | 9052985 |
Gene Symbol: RBMY1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006118.2 | NP_001006118.2 | 12 | 1617 | ||
XM_011531493.1 | XP_011529795.1 | 11 | 1506 | |||
XM_011531495.1 | XP_011529797.1 | 9 | 1284 | |||
XM_011531496.1 | XP_011529798.1 | 11 | 1456 | |||
XM_017030058.1 | XP_016885547.1 | 10 | 1322 |