Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USH1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297764.1 | NP_001284693.1 | 16 | 1421 | ||
NM_005709.3 | NP_005700.2 | 17 | 1478 | |||
NM_153676.3 | NP_710142.1 | 22 | 2378 | |||
NR_123738.1 | 16 | 1404 | ||||
XM_011519832.2 | XP_011518134.1 | 18 | 1631 | |||
XM_017017075.1 | XP_016872564.1 | 22 | 2378 | |||
GenBank mRNA | AB006955.1 | - | 16 | 1521 | ||
AB018687.1 | - | 17 | 1458 | |||
AF039699.1 | - | 16 | 1391 | |||
AF039700.1 | - | 17 | 1465 | |||
AK225614.1 | - | 17 | 1478 | |||
AK290788.1 | - | 17 | 1430 | |||
AK300936.1 | - | 17 | 1237 | |||
BC016057.1 | - | 16 | 1405 | 4867369 | ||
KU178498.1 | - | 16 | 1312 | |||
KU178499.1 | - | 17 | 1369 |