Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026793.1 | 2 | 552 | |||
GenBank mRNA | BC038537.1 | - | 2 | 552 |
Gene Symbol: LINC00202-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026795.1 | 4 | 3238 | |||
GenBank mRNA | AK097405.1 | - | 3 | 1405 | ||
AK128027.1 | - | 3 | 5605 |