Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR10G7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004463.1 | NP_001004463.1 | 1 | 763 |
Gene Symbol: OR10G8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004464.1 | NP_001004464.1 | 1 | 763 | ||
GenBank mRNA | BC136917.1 | - | 1 | 796 | 9020908 | |
BC136920.1 | - | 1 | 796 | 9020911 |