Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080487.2 | NP_001073956.2 | 5, 6 | 714 | ||
XM_017023230.1 | XP_016878719.1 | 7, 8 | 1969 | |||
GenBank mRNA | BC130002.1 | - | 4, 5 | 607 | ||
EU190484.1 | - | 5, 6 | 672 |