Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141936.2 | NP_001135408.2 | 3 | 316 | ||
NM_001168243.1 | NP_001161715.1 | 3 | 266 | |||
XM_011513469.2 | XP_011511771.1 | 4 | 1252 | |||
XM_017008190.1 | XP_016863679.1 | 4 | 325 | |||
GenBank mRNA | AI929674.1 | - | 3 | 260 | ||
BC038466.1 | - | 3 | 234 | |||
BC047038.1 | - | 3 | 316 | |||
BC062790.1 | - | 3 | 252 |