Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206922.2 | NP_996805.2 | 8 | 601 | ||
XM_005249103.3 | XP_005249160.1 | 7 | 2705 | |||
XM_011514609.2 | XP_011512911.1 | 7 | 2456 | |||
XM_017010856.1 | XP_016866345.1 | 6 | 2573 | |||
XM_017010857.1 | XP_016866346.1 | 5 | 2501 | |||
GenBank mRNA | AY555741.1 | - | 8 | 601 |