Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004067.3 | NP_001004067.1 | 31 | 3878 | ||
GenBank mRNA | BC006531.2 | - | 20 | 2499 | ||
BC013837.1 | - | 5 | 639 | |||
BQ948282.1 | - | 2 | 238 |
Gene Symbol: LOC102723728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720996.3 | XP_006721059.1 | 9 | 1123 |
Gene Symbol: NOMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014287.3 | NP_055102.3 | 31 | 3809 | ||
GenBank mRNA | AK125530.1 | - | 3 | 1461 | ||
BC065535.1 | - | 31 | 3762 | 6007603 | ||
X57398.1 | - | 31 | 3647 |
Gene Symbol: NOMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173614.2 | NP_775885.1 | 31 | 3815 | ||
GenBank mRNA | AK225803.1 | - | 31 | 3785 | ||
AL512687.1 | - | 8 | 1018 | |||
BC041131.1 | - | 31 | 3789 | 5757234 |