Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2Q2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243531.1 | NP_001230460.1 | 3 | 333 | ||
XM_017021854.1 | XP_016877343.1 | 3 | 893 | |||
XM_017021855.1 | XP_016877344.1 | 3 | 890 | |||
GenBank mRNA | CB050071.1 | - | 3 | 339 |