Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSFM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172695.1 | NP_001166166.1 | 5 | 557 | ||
NM_001172696.1 | NP_001166167.1 | 7 | 708 | |||
NM_005726.5 | NP_005717.3 | 6 | 645 | |||
GenBank mRNA | AF110399.1 | - | 6 | 591 | ||
AK295323.1 | - | 8 | 877 | |||
AK304621.1 | - | 5 | 541 | |||
AK313453.1 | - | 6 | 629 | |||
BC022862.1 | - | 7 | 681 | 4076880 | ||
L37936.1 | - | 5 | 496 |