Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002823.4 | NP_002814.3 | 2, 3 | 297 | ||
XM_011511519.1 | XP_011509821.1 | 2, 3 | 1027 | |||
XM_011511521.1 | XP_011509823.1 | 2, 3 | 926 | |||
GenBank mRNA | AF348514.1 | - | 2, 3 | 115 | ||
AF452640.1 | - | 2, 3 | 597 | |||
AK223605.1 | - | 2, 3 | 292 | |||
AK314106.1 | - | 2, 3 | 292 | |||
AY169282.1 | - | 2, 3 | 115 | |||
BC003510.1 | - | 2, 3 | 280 | 3610808 | ||
BC022433.1 | - | 2, 3 | 296 | 4277490 | ||
BC034921.1 | - | 2, 3 | 288 | 4335961 | ||
BC051265.1 | - | 2, 3 | 280 | 3832454 | ||
BC066905.1 | - | 2, 3 | 218 | 4830132 | ||
BC070480.1 | - | 2, 3 | 282 | 6205582 | ||
BC071647.1 | - | 2, 3 | 275 | 5112610 | ||
BC071879.1 | - | 2, 3 | 277 | 6425103 | ||
BC098420.1 | - | 2, 3 | 282 | 5017164 | ||
BM839980.1 | - | 2, 3 | 297 | |||
M14483.1 | - | 2, 3 | 247 | |||
M14794.1 | - | 2, 3 | 183 | |||
M26708.1 | - | 2, 3 | 270 |
Gene Symbol: LOC441454 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028350.1 | 1 | 1116 |
Gene Symbol: LOC728026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015408.1 | XP_016870897.1 | 1 | 771 |