Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALM2-AKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007203.4 | NP_009134.1 | 7, 8 | 766 | ||
NM_147150.2 | NP_671492.1 | 7, 8 | 766 | |||
GenBank mRNA | AB023137.1 | - | 7, 8 | 766 | ||
AJ303079.1 | - | 7, 8 | 754 | |||
AJ312217.1 | - | 8, 9 | 1316 | |||
BC140818.1 | - | 7, 8 | 607 | 9021686 | ||
BX649126.1 | - | 5, 6 | 552 |