Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F8A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007523.1 | NP_001007524.1 | 1 | 983 |
Gene Symbol: F8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012151.3 | NP_036283.2 | 1 | 998 | ||
GenBank mRNA | AL556125.3 | - | 1 | 992 | ||
BC039693.1 | - | 1 | 984 | 5214628 | ||
BC071963.1 | - | 1 | 988 | 5477018 |
Gene Symbol: F8A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007524.1 | NP_001007525.1 | 1 | 983 |