Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC93B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030930.3 | NP_112192.2 | 10 | 1460 | ||
XM_011545290.1 | XP_011543592.1 | 8 | 1070 | |||
XM_011545291.2 | XP_011543593.1 | 9 | 1352 | |||
GenBank mRNA | AJ271326.1 | - | 10 | 1419 | ||
AJ422142.1 | - | 10 | 1428 | |||
AY007125.1 | - | 9 | 1247 | |||
BC025669.1 | - | 10 | 1406 | |||
BC033623.1 | - | 10 | 1458 | |||
BC092472.1 | - | 10 | 1428 | 30348121 | ||
BC101568.1 | - | 10 | 1404 | 8069074 | ||
BC105104.1 | - | 10 | 1404 | 8144107 |