Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017011303.1 | XP_016866792.1 | 1 | 60 | ||
XM_017011304.1 | XP_016866793.1 | 1 | 60 |