Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177987.2 | NP_817124.1 | 4 | 1344 | ||
XM_011519459.2 | XP_011517761.1 | 4 | 1384 | |||
XM_011519460.2 | XP_011517762.1 | 3 | 1030 | |||
XM_017016192.1 | XP_016871681.1 | 2 | 1281 | |||
XM_017016193.1 | XP_016871682.1 | 2 | 1101 |
Gene Symbol: TUBB8P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC014971.1 | - | 3 | 1958 |
Gene Symbol: TUBBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027156.1 | 5 | 2500 |