Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282431.1 | NP_001269360.1 | 1 | 576 | ||
NM_005737.3 | NP_005728.2 | 1 | 572 | |||
GenBank mRNA | AB016811.1 | - | 1 | 80 | ||
AF493892.1 | - | 1 | 109 | |||
AJ579850.1 | - | 1 | 123 | |||
AJ579851.1 | - | 1 | 123 | |||
AK314119.1 | - | 1 | 113 | |||
BC089043.1 | - | 1 | 576 | 6578402 | ||
BT019378.1 | - | 1 | 109 | |||
BT019379.1 | - | 1 | 109 | |||
BU844902.1 | - | 1 | 576 | |||
Y17804.1 | - | 1 | 123 |