Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278632.1 | NP_001265561.1 | 6 | 2068 | ||
NM_001278633.1 | NP_001265562.1 | 7 | 2146 | |||
NM_001278634.1 | NP_001265563.1 | 6 | 2073 | |||
NM_001278635.1 | NP_001265564.1 | 7 | 2151 | |||
NM_016358.2 | NP_057442.1 | 5 | 1969 | |||
GenBank mRNA | AB690778.1 | - | 6 | 2068 | ||
AB690779.1 | - | 7 | 2146 | |||
AB690780.1 | - | 6 | 2073 | |||
AB690781.1 | - | 7 | 2151 | |||
BC110912.1 | - | 5 | 1857 | 3450161 |