Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005740.2 | NP_005731.1 | 4 | 403 | ||
GenBank mRNA | AK223349.1 | - | 4 | 390 | ||
AK290566.1 | - | 4 | 356 | |||
AL035366.1 | - | 4 | 396 | |||
AY453399.1 | - | 194 | ||||
BC002968.1 | - | 4 | 370 | 3543962 | ||
BT009847.1 | - | 3 | 175 | |||
BX473356.1 | - | 4 | 408 | |||
CR456487.1 | - | 3 | 260 | |||
CR542177.1 | - | 3 | 175 |