Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023037.2 | NP_075463.2 | 29, 30 | 4251 | ||
XM_006719749.3 | XP_006719812.1 | 29, 30 | 5332 | |||
XM_017020303.1 | XP_016875792.1 | 29, 30 | 5809 | |||
XM_017020304.1 | XP_016875793.1 | 29, 30 | 4251 | |||
XM_017020305.1 | XP_016875794.1 | 29, 30 | 5828 | |||
XM_017020306.1 | XP_016875795.1 | 8, 9 | 970 | |||
XM_017020307.1 | XP_016875796.1 | 2, 3 | 323 | |||
GenBank mRNA | AL049784.1 | - | 29, 30 | 4251 | ||
U50534.1 | - | 29, 30 | 4250 |