Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008211.1 | NP_001008212.1 | 7, 8 | 1089 | ||
NM_001008212.1 | NP_001008213.1 | 6, 7 | 1020 | |||
NM_001008213.1 | NP_001008214.1 | 7, 8 | 1074 | |||
NM_021980.4 | NP_068815.2 | 5, 6 | 868 | |||
GenBank mRNA | AF420371.1 | - | 7, 8 | 946 | ||
AF420372.1 | - | 5, 6 | 725 | |||
AF420373.1 | - | 6, 7 | 877 | |||
AK055403.1 | - | 6, 7 | 1309 | |||
AK303715.1 | - | 5, 6 | 886 | |||
AK316171.1 | - | 5, 6 | 908 | |||
BC013876.2 | - | 5, 6 | 781 | 3831267 | ||
KU178502.1 | - | 4, 5 | 636 | |||
KU178503.1 | - | 4, 5 | 636 |