Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMF1-BGLAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199661.1 | NP_001186590.1 | 5, 6 | 565 | ||
NM_001199662.1 | NP_001186591.1 | 5, 6 | 626 | |||
NM_001199663.1 | NP_001186592.1 | 4, 5 | 430 | |||
NM_001199664.1 | NP_001186593.1 | 3, 4 | 419 | |||
GenBank mRNA | BC033656.2 | - | 4, 5 | 420 | ||
CD049192.1 | - | 5, 6 | 634 | |||
CD388852.1 | - | 5, 6 | 565 | |||
FY213424.1 | - | 3, 4 | 332 |
Gene Symbol: BGLAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199173.5 | NP_954642.1 | 2, 3 | 368 | ||
GenBank mRNA | BC113432.1 | - | 2, 3 | 170 | 8322484 | |
BC113434.1 | - | 2, 3 | 170 | 8322486 | ||
X51699.1 | - | 2, 3 | 116 | |||
X53698.1 | - | 2, 3 | 116 |